missing translation for 'onlineSavingsMsg'
Learn More
Gallium ingot, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11301238

Gallium ingot, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11301238
50 g
Click to view available options
Quantity:
50 g
500 g
This item is not returnable. View return policy

Product Code. 11301238

Brand: Thermo Scientific Alfa Aesar 010185.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Requires overnight shipping with cold pack

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-55-3
Molecular Formula Ga
Molecular Weight (g/mol) 69.72
MDL Number MFCD00134045
InChI Key GYHNNYVSQQEPJS-UHFFFAOYSA-N
Synonym gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atomShow MoreShow Less
PubChem CID 5360835
ChEBI CHEBI:49631
IUPAC Name gallium
SMILES [Ga]

Specifications

Color Silver to Gray
Appearance Silver Gray ingot
Assay Percent Range 99.9999% (Metals basis)
Quantity 50 g
Form Ingot
Chemical Name or Material Gallium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX18K060 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number010185.18
Lot NumberW19K033 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog Number010185.18
Lot NumberQ03K051 Certificate TypeCertificate of Analysis Date10/04/2024 Catalog Number010185.18
Lot NumberR12J043 Certificate TypeCertificate of Analysis Date06/06/2023 Catalog Number010185.18
Lot NumberT25I044 Certificate TypeCertificate of Analysis Date30/08/2022 Catalog Number010186.18,010185.18,010185.14,010185.36,010185.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gallium ingot, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.