missing translation for 'onlineSavingsMsg'
Learn More

Gallium metal, packaged in polyethylene bottle, 99.9% (metals basis)

Product Code. 15487095
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 15487095

Brand: Thermo Scientific Alfa Aesar 045841.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

May be solid or liquid due to ambient temperature | CAS: 7440-55-3 | Ga | 69.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-55-3
Molecular Formula Ga
Molecular Weight (g/mol) 69.72
MDL Number MFCD00134045
InChI Key GYHNNYVSQQEPJS-UHFFFAOYSA-N
Synonym gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
PubChem CID 5360835
ChEBI CHEBI:49631
IUPAC Name gallium
SMILES [Ga]

Specifications

Color Silver to Gray
Appearance Silver Gray
Quantity 500 g
Assay Percent Range 99.9999% (Metals basis)
Form Solid and/or Liquid
Chemical Name or Material Gallium
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P234-Keep only in original container.
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P390-Absorb spillage to prevent material damage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.