Learn More
Gallium pellets, 99.9999% (metals basis), Thermo Scientific Chemicals
£103.00 - £1749.71
Identificadores de productos químicos
CAS | 7440-55-3 |
---|---|
Molecular Formula | Ga |
Molecular Weight (g/mol) | 69.72 |
MDL Number | MFCD00134045 |
InChI Key | GYHNNYVSQQEPJS-UHFFFAOYSA-N |
Synonym | gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom |
PubChem CID | 5360835 |
ChEBI | CHEBI:49631 |
IUPAC Name | gallium |
SMILES | [Ga] |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
11408440
|
Thermo Scientific Alfa Aesar
044324.09 |
10 g | N/A | ||||||
11498430
|
Thermo Scientific Alfa Aesar
044324.18 |
50 g |
£402.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11418440
|
Thermo Scientific Alfa Aesar
044324.30 |
250 g |
£1749.71
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
7440-55-3 | |
69.72 | |
GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
5360835 | |
gallium |
Ga | |
MFCD00134045 | |
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom | |
CHEBI:49631 | |
[Ga] |
Especificaciones
7440-55-3 | |
Ga | |
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom | |
[Ga] | |
69.72 | |
CHEBI:49631 | |
Gallium |
99.9999%, metal basis | |
MFCD00134045 | |
GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
gallium | |
5360835 | |
Pellet |
Seguridad y manipulación
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
missing translation for 'dotInformation' : DOT Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 231-163-8
missing translation for 'rtecsNumber' : LW8600000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only