missing translation for 'onlineSavingsMsg'
Learn More

Gallium Tin alloy, 99.99% (metals basis)

Product Code. p-4781905
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 11303427

Brand: Thermo Scientific Alfa Aesar 018161.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7440-31-5 | Sn | 118.71 g/mol

Gallium Tin alloy is used to cool computer chips in place of water.  Used in medical thermometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-31-5, 7440-55-3
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Color Silver
Melting Point ≥25°C
Appearance Silver-colored liquid
Quantity 1 g
Assay Percent Range 99.99% (metals basis)
Form Melting Solid
Solubility Information Insoluble in water.
Concentration or Composition (by Analyte or Components) Tin: 8.0%; Gallium: 92.0%
Chemical Name or Material Gallium Tin alloy

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.