missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Galus S3 Duo Bond Boot

Product Code. 17542476
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 17542476

Brand: Emma Safety Footwear MM58086644

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Insulated with fine quality sheepskin, there is no warmer shoe than the Galus to wear in the coldest of conditions.

  • A naturally better boot.
  • Even in this age of hi-tec, high performance synthetics, sometimes nature's materials are still the best. Which is why we've insulated the Galus with fine quality sheepskin. There's no warmer shoe to wear in the coldest of conditions. And if the temperature suddenly rises up to 300 degrees on the outside, it's reassuring to know the nitrile rubber outsole won't yield one bit.
  • Industries:
    • Chemical industry
    • Construction & technology
  • Safety shoe
    • S3-Safetyshoe with sole plate for wet work

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Zipper
Color Black
For Use With (Application) Chemical Industry, Construction and Technology
Liner Material Sheepskin
Material Leather
Size 44
Sole Features GripForce™ Duo
Sole Material Nitrile Rubber
Toe Material Steel
Fit Extra-wide Width
EN ISO 20345 S3
Product Type Safety Boot
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.