Learn More
Emma Safety Footwear Galus S3 Duo Bond Boot
Insulated with fine quality sheepskin, there is no warmer shoe than the Galus to wear in the coldest of conditions.
Brand:  Emma Safety Footwear MM58086639
 View more versions of this product
Product Code. 17592466
  • £148.00 / 1 pair
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A naturally better boot.
  • Even in this age of hi-tec, high performance synthetics, sometimes nature's materials are still the best. Which is why we've insulated the Galus with fine quality sheepskin. There's no warmer shoe to wear in the coldest of conditions. And if the temperature suddenly rises up to 300 degrees on the outside, it's reassuring to know the nitrile rubber outsole won't yield one bit.
  • Industries:
    • Chemical industry
    • Construction & technology
  • Safety shoe
    • S3-Safetyshoe with sole plate for wet work
Specifications
Specifications
ISO 20345:2011
Black
Sheepskin
39
Nitrile Rubber
Extra-wide Width
Safety Boot
Zipper
Chemical Industry, Construction and Technology
Leather
GripForce™ Duo
Steel
S3
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Brochures
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Emma Safety Footwear Galus S3 Duo Bond Boot > Safety Boot; Size: 39