Learn More
WTW™ Galvanic Dissolved Oxygen Sensor for WTW Conductivity and Multiparameter
proven conventional sensors for pH, ORP, ISE, conductivity and dissolved oxygen are the masters of all measurements in the field and in the lab
Brand:  WTW™ 201425
 View more versions of this product
Product Code. 10160691
  • £1215.00 / Each
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The CellOx™ 325 is a dissolved oxygen sensor with integrated temperature compensation and membrane leak monitoring, which can be used in the lab as well as in the field. Its features include a quick response time (t99<60 s) and low-maintenance operation. Its fields of use include surface waters, measurements in wastewater treatment plants, BOD directions, but also industrial and other applications. It is filled with electrolyte and achieves a long dwelling time up to 6 months. The calibration vessel is part of the delivery.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
StirrOx™ G Self-stirring Dissolved Oxygen Sensor
20 m
20m L Cable Cleaning solution, abrasive foil, 3 exchange membrane heads and electrolyte
Conductivity and multiparameter meters
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products