missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Radiation Safety Storage Bins
Radiation safety bins that are ideal for protection against Beta and Gamma radiation.
Brand: Fisherbrand™ CSR-B53G
This item is not returnable.
View return policy
Description
- Gamma - 53L, use with bag CSR-BAG5 40 x 49 x 27cm
- Supplied in small, medium and large sizes and with curved base 15cm-deep for use with safety trays or flat 30x30cm base for under-the-bench protection
- A range of angles offers increased maneuverability, while clear optical acrylic aids visualization
- Provides clear, non-distorted vision, reducing light images and shadows, especially when the user is in a sitting position
- Available in two sizes, with either a flat base to provide bench protection, or a curved base which fits conveniently over safety trays
Specifications
For storing radioactive materials | |
424mm | |
Radiation Bin | |
514mm |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Radiation Safety Storage Bins > Capacity: 53L
Spot an opportunity for improvement?Share a Content Correction