missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gamma cystathionase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12217-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human CTH.
This gene encodes a cytoplasmic enzyme in the trans-sulfuration pathway that converts cystathione derived from methionine into cysteine. Glutathione synthesis in the liver is dependent upon the availability of cysteine. Mutations in this gene cause cystathioninuria. Alternative splicing of this gene results in two transcript variants encoding different isoforms.Specifications
Gamma cystathionase | |
Polyclonal | |
Unconjugated | |
CTH | |
CSE, CTH, Cystathionine gamma lyase, Gamma cystathionase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
107869, 1491, 24962 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P18757, P32929, Q8VCN5 | |
Cth | |
Gamma cystathionase Fusion Protein Ag2872 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |