Learn More
Keystone Cleanroom Products Gamma-Irradiated Retractable Ultra Fine Point Sharpie
Ready-to-use gamma irradiated pens and markers eliminate the risks associated with surface sanitization procedures.
Brand: Keystone Cleanroom Products S01RETMARKBLUE
Additional Details : Weight : 8.98110kg
Description
- Gamma irradiated
- Lot controlled
- Ready to use
- Packaged in easy to open Tyvek™ peel pouches
- Minimize contamination risks
- Simplifies conformance to SOP's
- Eliminate the need for wiping down contaminated writing instruments
Specifications
Plastic | |
Not Autoclavable | |
Yes |
Blue | |
COC, Certificate of Irradiation | |
Sterile |