Learn More
Fisherbrand™ Radiation Storage Boxes
Ideal for protection against Beta or Gamma radiation, these Gamma Midi Duo Radiation boxes with hinged lid and central swivel covers are used in many research, scientific, and other laboratory environments.
Brand:  Fisherbrand™ CSR-BDUOG
Product Code. 12337137
  • £876.00 / Each
Estimated Shipment: 08-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Gamma Midi Duo 2 Pig-Pen Box 7 x 10 x6cm
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • Also supplied is range of floor-standing and benchtop bins with anti-slip feet and hinged lids
  • These serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, while the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
Specifications
Specifications
To hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
60mm
Microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
Radiation Storage Box
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Radiation Storage Boxes > Height: 60mm