Learn More
Fisherbrand™ Gamma Radiation Safe Midi Storage Box
Provides radiation protection shielding for use in laboratories.
Brand:  Fisherbrand™ CSR-B0.8G
Product Code. 12327107
  • £1760.00 / Each
Estimated Shipment: 06-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured with hinged lids
  • Transparent shielding
  • Serves as solution for short-term storage of radioactive waste or radioisotopes
  • Constructed of acrylic provides easy fabrication and manufacturability
Specifications
Specifications
16.5 x 8.5 x 6cm (6.5 x 3.35 x 2.36 in.)
Radioisotopes or radioactive waste
Midi Storage Box
Gamma radiation shielded
Hinged lid
Acrylic
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Gamma Radiation Safe Midi Storage Box > Gamma Radiation Safe Midi Storage Box