missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Beta Radiation Safe Storage Bin
Provides radiation protection shielding for use in laboratories.
Brand: Fisherbrand™ CSR-B2MCTIP
This item is not returnable.
View return policy
Description
- Serves as solution for short-term storage of radioactive waste or radioisotopes
Specifications
Beta radiation safe | |
Bag dimension: L x W x H: 13 x 13 x 13cm (5.12 x 5.12 x 5.12 in.) | |
Storage bin | |
2 L |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Beta Radiation Safe Storage Bin > Capacity: 2L; Use with bag dimension L x W x H: 13 x 13 x 13cm
Spot an opportunity for improvement?Share a Content Correction