Learn More
Thermo Scientific™ gamma-Secretase Inhibitor II
Brand:  Thermo Scientific™ J64828.#0
 View more versions of this product
Product Code. 15476599
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 mg
-30°C to -10°C
C20H30N2O4
MW167
Solid
Off-white
A cell-permeable, reversible and selective peptidomimetic inhibitor of --secretase
362.47
gamma-Secretase Inhibitor