Learn More
Cytiva GammaBind™ G Sepharose™ Medium
Use recombinant GammaBind™ for laboratory scale antibody purification. Cytiva Life Sciences™ GammaBind™ G Sepharose™ Medium binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse
Brand: Cytiva 17-0885-02
Code : GE
Additional Details : CAS Number : 79-06-1 Weight : 0.05200kg
Description
Binds to all IgG subclasses from human, mouse, and rat and total IgG from guinea pig, rabbit, goat, cow, sheep, and horse. Engineered for antibody purification at laboratory scale.
- Will not cross-react with other serum proteins such as IgM, IgE, IgA or transferrin
- Ligand: GammaBind G type 2 lacking albumin- binding region
- Ligand coupling method: cyanogen bromide activation
- Ligand density: ≈ 3mg GammaBind G, type 2/mL medium
- Number of IgG binding sites per ligand: 2
- Matrix: Agarose, 4%
- Average particle size: 90μm
- Binding capacity: >18 mg human IgG/ml, 6 mg mouse IgG/ml
- pH stability: 3-9 (working), 2-9 (short term)
Specifications
Sepharose Medium | |
≈3 mg GammaBind G, Type 2/mL drained gel | |
3 to 9 | |
For protein purification by affinity coupling | |
25 mL |
>18 mg/mL Human IgG, 6 mg/mL Mouse IgG | |
45 to 165 μm | |
25 mL | |
PBS pH 7, 20% Ethanol |
Safety and Handling
- GammaBind G Sepharose, 25 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.