missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GammaBind™ Plus Sepharose™ Medium
Engineered for better binding to rat and mouse IgG subclasses
Brand: Cytiva 17-0886-01
This item is not returnable.
View return policy
Description
- Recombinant Protein G type 3 coupled to Sepharose CL-6B for easy binding and fast separation
- More enhanced binding capabilities than GammaBind G at both laboratory and process scale
- Yields up to 18mg/mL mouse IgG
- Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse
Specifications
GammaBind Plus Sepharose | |
90 μm | |
5 mL |
Recombinant GammaBind G type 3 lacking albumin-binding region | |
2°C to 8°C, 20% Ethanol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- gammabind plus sepharose
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction