Learn More
Ansell™ Gammex™ PF Powder-Free Latex Surgical Gloves
Provides an optimal combination of safety, based on Ansell's powder-free technology, with paralleled levels of comfort and ease-of-use. Ansell Edmont™ Gammex™ PF Powder-Free Latex Surgical Gloves have a very high tear resistance, with a favorable strength-to-thickness ratio for optimal barrier protection.
Brand:  Ansell™ AJ330048075
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 17131985
  • £190.80 / 50 pairs
Estimated Shipment: 14-08-2024
to see stock.
Add to basket
Description
Description
  • Micro-rough outer surface provides a secure grip in both wet and dry conditions
  • Synthetic inner coating reduces possible contact with latex allergens
  • Resistant to alcohol-based hand disinfectants and viral penetration
  • Delivers elasticity, sensitivity and comfort plus a natural fit that helps prevent hand fatigue
Specifications
Specifications
Single-use
No
Powder Free
Straight
Tears, alcohol-based hand disinfectants, viral penetration
Ambidextrous
Latex
7.5
50 Pairs/Box