missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAP-43 Rabbit anti-Human, Mouse, Rat, Clone: 8E7C8, Novus Biologicals™
Rabbit Monoclonal Antibody
£154.00 - £384.00
Specifications
Antigen | GAP-43 |
---|---|
Clone | 8E7C8 |
Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18318854
|
Bio-Techne
NBP3-15636-20UL |
20 μg |
£154.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18346835
|
Bio-Techne
NBP3-15636-100UL |
100 μg |
£384.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GAP-43 Monoclonal antibody specifically detects GAP-43 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
GAP-43 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1 | |
A synthetic peptide corresponding to a sequence within amino acids 139-238 of human GAP-43 (P17677). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
8E7C8 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2596 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title