missing translation for 'onlineSavingsMsg'
Learn More

GAPDH Mouse anti-Human, Mouse, Plant, Rat, Zebrafish, HRP, Clone: 1E6D9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech HRP-60004-100UL

Product Code. 16891236

  • £189.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is raised against full length GAPDH of human origin. It can recognize the 36 kDa GAPDH protein in most cells/tissues. In addition, a band below 36 kDa can always be detected as the isoform or spliced product of GAPDH. Please note that some p

GAPDH (Glyceraldehyde-3-phosphate dehydrogenase) is a catalytic enzyme commonly known to be involved in glycolysis. GAPDH exists as a tetramer of identical 37-kDa subunits and catalyzes the reversible reduction of 1,3-bisphosphoglycerate to glyceraldehyde 3-phosphophate in the presence of NADPH. Apart from playing a key role in glycolysis, GAPDH is ubiquitously expressed and displays other activities unrelated to its glycolytic function. GAPDH is reported to be involved in the processes of DNA replication, DNA repair, nuclear RNA export, membrane fusion and microtubule bundling. Studies provide evidence of GAPDH playing an essential part in gene expression observed in apoptosis and as part of the cellular phenotype of age-related neurodegenerative diseases. Further, GAPDH is involved in other cellular processes ranging from membrane fusion, and neuronal apoptosis in cancer. GAPDH is reported to bind to a variety of other proteins, including the amyloid precursor protein, mutations in which cause some forms of Alzheimer′s disease (AD), and the polyglutamine tracts of Huntingtin, the protein product aberrant forms of which are causative of Huntington′s disease. Associations between GAPDH, actin and tubulin have also be reported. Since GAPDH is expressed at high levels in most tissues, it is useful as protein loading control in Western Blot analysis.
Specifications

Specifications

GAPDH
Monoclonal
0.67 mg/mL
PBS with 50% glycerol and 0.02% thimerosal; pH 7.3
P04406, P04797, P16858, Q5XJ10
gapdh
Antigen Affinity Chromatography
RUO
14433, 24383, 2597, 317743
-20°C
Liquid
Western Blot
1E6D9
HRP
GAPDH
G3PD, GAPD, GAPDH, HRP-G3PD, HRP-GAPD, HRP-GAPDH, HRP-OK/SW cl.12, OK/SW cl.12
Mouse
100 μL
Primary
Human, Mouse, Plant, Rat, Zebrafish
Antibody
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.