Learn More
Thermo Scientific™ Gas Chromatography 3-Port SilFlow Device
Switch your GC without the need to vent using the Thermo Scientific™ 3-Port SilFlow™ Device.
Brand: Thermo Scientific™ 60201-398
Code : NEW
Additional Details : Weight : 0.20000kg
Description
Switch your GC without the need to vent using the Thermo Scientific™ 3-Port SilFlow™ Device. This device comprises a gas module with microfluidics and Thermo Scientific™ SilTite™ FingerTite fittings for easy set up and a reliable seal.
Specifications
3-Port SilFlow MCD | |
3-Port SilFlow MCD (0.25/0.32) | |
1/Ea. | |
Gas Chromatography Systems |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.