missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Gas Cylinder Lockout

Safely lockout gas cylinders to help avoid workplace accidents. The Brady™ Gas Cylinder Lockout is adjustable and installs in seconds. There is no need to match your lockout device to varying diameters or threads of cylinder caps.

Brand:  Brady™ 045629

Product Code. 11711855

  • £53.05 / Each
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • No need to match your lockout device to varying diameters and threads of cylinder caps
  • Adjustable and installs in just seconds
  • Device is manufactured to accommodate rings up to 3.5 in. diameter
Specifications

Specifications

ASSY, GAS CAP CYLINDER LO
Polypropylene
1 Ea.
Red
158.80 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Gas Cylinder Lockout >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.