Learn More
BW Technologies™ Gas Detectors
Brand:  BW Technologies™ M5IR-XWBY-R-D-D-Y-E-00
 View more versions of this product
Product Code. 11960917
  • £3260.00 / Each
Explore more special offers
Specifications
Audible
2 AA
38 mm
Gas Detector
IP67
370g
2 Years
Ex II 1 G T4 Eex ia IIC T4 except CO2 detector
Oxygen, Carbon Monoxide, Hydrogen Sulfide, Sulfur Dioxide, VOC, Carbon Dioxide Gas and Fuel
145 mm
74 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
BW Technologies™ Gas Detectors > Detects: 5 Gas: O2, CO, H2S, SO2 / VOC / C02 gas and fuel