Learn More
Trajan Scientific and Medical Gas Filter Connecting Unit
The gas filter connecting unit has inlet and outlet connectors for the gas line and the system can be wall-mounted or fixed to a bench. Connecting units come with a 1, 2 or 4 filter capacity and are available for 1/4 in. or 1/8 in. gas lines
Brand:  Trajan Scientific and Medical 1035024
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15760959
  • £413.00 / Each
Estimated Shipment: 22-08-2024
to see stock.
Description
Description
  • Easily installed into the gas line
  • Can be either wall-mounted or fixed to a bench
  • Available with a 1, 2 or 4 filter capacity
Specifications
Specifications
1/4 in. fittings
Gas filters
1/4 in.
2-position Connecting Unit