Learn More
Trajan Scientific and Medical Gas Filter - Hydrocarbon
Prevent contaminants in gases from significantly impacting the quality of results. Oxygen, hydrocarbons and moisture can lead to problems such as noisy baselines, moisture entering the GC column, excessive bleed and septa degradation.
Brand:  Trajan Scientific and Medical 1035230
Additional Details : Weight : 2.00000kg
Product Code. 15760969
  • £157.00 / Each
Estimated Shipment: 08-08-2024
to see stock.
Description
Description
  • Capacity of approximately 7 g organics dpending upon impurities, for long filter lifetime
  • Can be used in combination with other filters through a simple connecting unit
Specifications
Specifications
Approx. 7 g, depending upon impurities
Hydrocarbon
Connecting unit
No
Removes organic compounds from gas streams