missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical Gas Filter Kit - Carrier Gas

Prevent contaminants in gases from significantly impacting the quality of results. Oxygen, hydrocarbons and moisture can lead to problems such as noisy baselines, moisture entering the GC column, excessive bleed and septa degradation.

Brand:  Trajan Scientific and Medical 1035158

Additional Details : Weight : 10.00000kg

 View more versions of this product

Product Code. 15710969

  • £304.50 / Each
Estimated Shipment: 22-08-2024
to see stock.

Description

Description

  • Capacity of 100 mL oxygen, 1g water, organics depending upon impurities
  • Indicator color change tells you when to change your filter
  • Can be used in combination with other filters through a simple connecting unit
  • Kit consists of gas filter and connecting unit
Specifications

Specifications

100 mL
Carrier Gas
1 x 1-position connecting unit, 1 x gas filter (carrier gas)
1/8 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For research use only