Learn More
Trajan Scientific and Medical Gas Filter - Moisture
Prevent contaminants in gases from significantly impacting the quality of results. Oxygen, hydrocarbons and moisture can lead to problems such as noisy baselines, moisture entering the GC column, excessive bleed and septa degradation.
Brand:  Trajan Scientific and Medical 1035220
Additional Details : Weight : 2.00000kg
Product Code. 15750969
  • £187.00 / Each
Estimated Shipment: 07-08-2024
to see stock.
Description
Description
  • Capacity of 7.2 g water for long filter lifetime
  • Indicator color change tells you when to change your filter
  • Can be used in combination with other filters through a simple connecting unit
Specifications
Specifications
7.2 g H2O
Moisture
Connecting unit
From green to pale brown
Removes water, oil and other foreign material from carrier gas