Learn More
Cole-Parmer™ Gas Regulator
Brand:  Cole-Parmer™ 68826-36
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15126754
  • £76.80 / Each
Estimated Shipment: 28-08-2024
to see stock.
Specifications
Sub-miniature Regulator 0 To 5 psig; 1/16 in. NPT(F)
Cylinder
29.46 mm
Gas Regulator
Brass