Learn More
Spectrum Labs™ Gas Safety Stand
Product Code. 15381002
Click to view available options
Product Type:
Gas cylinder safety stand
Unit Size:
Each
Product Code. 15381002
Brand: Spectrum Labs™ 148 702
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Spectrum Labs™ Gas Safety Stand locks cylinders into place to keep them from falling.
  • Keeps large cylinders securely in place
  • Locks for maximum safety
Specifications
Product Type Gas cylinder safety stand
For Use With (Application) Large cylinders, 18 to 25 cm in diameter