missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Gas Sampling Tubes

Product Code. p-7222489
Click to view available options
Capacity (Metric):
150 mL
350 mL
1000 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10201901

Brand: Lenz Laborglasintrumente 05680070

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

  • Gas sampling tubes acc. to DIN 12473-1
  • Available with or without nozzle (with thread GL14 and septa)
  • With two one-way stopcocks NS 14 (with retaining device)
  • Connections: capillary tube (outer dia. 8 mm, inner dia. 2,7 mm)
  • Glass olive (outer dia. 9 mm)
  • With inscription label
  • Made of DURAN™ tubing

Specifications

Capacity (Metric) 1000 mL
Material Duran Glass
Includes Without nozzle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.