Learn More
Trajan™ Gas-Tight Autosampler Syringes
Replace syringes for Agilent™ Technologies, Varian™, Perkin Elmer™, Shimadzu™, Carlo Erba™, Fisons™, CTC/Leap™, Hitachi™, and more
£84.60 - £128.00
Specifications
Diameter (English) Outer Needle | 0.02 in. |
---|---|
Product Type | Autosampler Syringe |
Quantity | Ea. |
Product Code | Brand | Connection Type | Volume (Metric) | Needle Gauge | For Use With (Equipment) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connection Type | Volume (Metric) | Needle Gauge | For Use With (Equipment) | Price | Quantity & Availability | ||
12997556
|
Trajan™
005330 |
Removable | 100 μL | 22 G | Carlo Erba/Fisons/Leap/CTC ASD200 and AS800 |
£84.60
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10742002
|
Trajan™
001957 |
Fixed | 5 μL | 23 G | PerkinElmer Autosystem |
£128.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12977556
|
Trajan™
005331 |
Fixed | 100 μL | 22 G | CTC, Carlo Erba, Fisons Autosampler |
£84.60
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Syringe needles are cone point unless otherwise noted.Order Info
Additional syringes are available for other autosamplers made by the manufacturers represented here, as well as syringes for Unicam™, Waters™, and Siemens™ instruments. For information, contact your Fisher Scientific Customer Service Representative.
Specifications
0.02 in. | |
Autosampler Syringe | |
Ea. |