Learn More
Trajan Scientific and Medical™ Gas Tight Fixed Needle Autosampler Syringe, Cone Tip
Designed to provide improved solvent resistance, greater temperature range and increased operational smoothness.
£94.80 - £187.00
Specifications
Material | PTFE-tipped (Plunger) |
---|---|
Product Type | Gastight |
Needle Point Style | Cone |
Connection Type | Fixed |
Product Code | Brand | Volume (Metric) | Diameter (Metric) Inner Needle | Diameter (Metric) Outer Needle | Length (Metric) Needle | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Diameter (Metric) Inner Needle | Diameter (Metric) Outer Needle | Length (Metric) Needle | Price | Quantity & Availability | ||
13126105
|
Trajan Scientific and Medical™
007862 |
500 mL | 0.32 mm | 0.63 mm | 57 mm |
£116.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13136105
|
Trajan Scientific and Medical™
007864 |
500 μL | 0.32 mm | 0.63 mm | 85 mm |
£137.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13139215
|
Trajan Scientific and Medical™
005862 |
100 μL | 0.11 mm | 0.63 mm | 57 mm |
£94.80
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13146105
|
Trajan Scientific and Medical™
007866 |
500 μL | 0.20 mm | 0.47 mm | 57 mm |
£164.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13149215
|
Trajan Scientific and Medical™
005864 |
100 μL | 0.11 mm | 0.63 mm | 85 mm |
£122.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13156105
|
Trajan Scientific and Medical™
007868 |
500 μL | 0.20 mm | 0.47 mm | 85 mm |
£187.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13159215
|
Trajan Scientific and Medical™
005866 |
100 μL | 0.11 mm | 0.47 mm | 57 mm |
£94.80
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13166095
|
Trajan Scientific and Medical™
006862 |
250 μL | 0.32 mm | 0.63 mm | 57 mm |
£102.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13169215
|
Trajan Scientific and Medical™
005868 |
100 μL | 0.11 mm | 0.47 mm | 85 mm |
£122.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13176095
|
Trajan Scientific and Medical™
006864 |
250 μL | 0.32 mm | 0.63 mm | 85 mm |
£130.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13186095
|
Trajan Scientific and Medical™
006866 |
250 μL | 0.20 mm | 0.47 mm | 57 mm |
£102.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13196095
|
Trajan Scientific and Medical™
006868 |
250 μL | 0.20 mm | 0.47 mm | 85 mm |
£130.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Thermo Scientific instruments
Specifications
PTFE-tipped (Plunger) | |
Gastight | |
Cone | |
Fixed |