missing translation for 'onlineSavingsMsg'
Learn More

3M™ Gas and Vapour Filters

3M™ Gas and Vapour Filters; reusable masks with a bayonet connection and are designed to optimise your field of vision. Our filters offer lightweight and well-balanced breathing protection for hazardous environments.

Brand:  3M™ 7100006000

45.40 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15729067

  • £127.00 / 4 pairs
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Colour-coded labels according to the EN14387 classification system to show the type of protection they provide.These lightweight filters combine protection with comfort, and they’re compatible with our range of half and full-face masks for safe working in hazardous environments. They offer low breathing resistance and good balance when connected to your mask, and they’re designed to optimise your field of vision.They can be clicked into place quickly and easily, and they can be used with any 3M™ Reusable Masks that feature a bayonet-style connection. They can also be used in combination with 3M™ Particulate Filters and 3M™ 501 Particle Filter Retainers.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cartridge Filter
Bayonet
White
Gas and Vapor Cartridge
Acid cleaning, Metal pickling, Metal etching and use, Chemical manufacture and handling
6057
100 g
CE
Inorganic vapors and acid gases, Organic Vapors of boiling point >65°C
Combination Acid Gas/Organic Vapor
ABE1
Half and Full Facemasks
8/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers