missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Gas and Vapour Filters
3M™ Gas and Vapour Filters; reusable masks with a bayonet connection and are designed to optimise your field of vision. Our filters offer lightweight and well-balanced breathing protection for hazardous environments.
Brand: 3M™ 7000030099
This item is not returnable.
View return policy
Description
Colour-coded labels according to the EN14387 classification system to show the type of protection they provide.These lightweight filters combine protection with comfort, and they’re compatible with our range of half and full-face masks for safe working in hazardous environments. They offer low breathing resistance and good balance when connected to your mask, and they’re designed to optimise your field of vision.They can be clicked into place quickly and easily, and they can be used with any 3M™ Reusable Masks that feature a bayonet-style connection. They can also be used in combination with 3M™ Particulate Filters and 3M™ 501 Particle Filter Retainers.
Specifications
Organic Vapor | |
Bayonet | |
Gas, Vapor and Particulate Filter | |
Typical industrial applications for filters are laboratories and particulate applications | |
6099 |
EN14387:2004 + A1:2008, CE Approved | |
Organic gases and vapors (boiling point above 65°C) and particles, organic vapors (boiling point above 65°C), acidic gases, mercury and particles, organic vapors (boiling point above 65°C), inorganic vapors, acidic gases, ammonia and its derivatives, formaldehyde up to 10 ppm and particles, organic vapors (boiling point above 65°C), inorganic vapors, acid gases, ammonia and its derivatives, mercury, formaldehyde up to 10 ppm and particles, organic vapors, mercury vapors and particles, from one Component consisting of organic vapors (boiling point below 65°C) and particles | |
ABEK2P3R | |
Half and Full Facemasks | |
4/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction