Learn More
3M™ Gas and Vapour Filters
3M™ Gas and Vapour Filters; reusable masks with a bayonet connection and are designed to optimise your field of vision. Our filters offer lightweight and well-balanced breathing protection for hazardous environments.
Brand: 3M™ 7000034746
44.34 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Colour-coded labels according to the EN14387 classification system to show the type of protection they provide.These lightweight filters combine protection with comfort, and they’re compatible with our range of half and full-face masks for safe working in hazardous environments. They offer low breathing resistance and good balance when connected to your mask, and they’re designed to optimise your field of vision.They can be clicked into place quickly and easily, and they can be used with any 3M™ Reusable Masks that feature a bayonet-style connection. They can also be used in combination with 3M™ Particulate Filters and 3M™ 501 Particle Filter Retainers.
Specifications
Cartridge Filter | |
Bayonet | |
White | |
Gas and Vapor Cartridge | |
Protect Against Organic Vapors | |
6075 | |
100 g |
CE | |
Inorganic vapors and acid gases, Organic vapors bp of >65°C | |
Formaldehyde/Organic Vapor | |
A1 | |
Half and Full Facemasks | |
1 Pair |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.