missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Gas and Vapour Filters
Click to view available options
Model:
6038
6051
6054
6055
6057
6059
6075
6099
Unit Size:
10 pairs
2 pairs
32 pairs
4 pairs
Description
Colour-coded labels according to the EN14387 classification system to show the type of protection they provide.These lightweight filters combine protection with comfort, and they’re compatible with our range of half and full-face masks for safe working in hazardous environments. They offer low breathing resistance and good balance when connected to your mask, and they’re designed to optimise your field of vision.They can be clicked into place quickly and easily, and they can be used with any 3M™ Reusable Masks that feature a bayonet-style connection. They can also be used in combination with 3M™ Particulate Filters and 3M™ 501 Particle Filter Retainers.
Specifications
Specifications
| Certifications/Compliance | EN14387:2004 + A1:2008 |
| Connection Type | Bayonet |
| Description | Organic vapors bp >65°C |
| Filter Type | Organic Vapor (OV) |
| Product Type | Gas and Vapor Cartridge |
| Filter Efficiency | A1 |
| For Use With (Application) | Anywhere conventional paints are used (subject to usage conditions), Vehicle manufacture, Aircraft manufacture & Refurbishment, Ink & Dye manufacturer and use, Adhesive manufacturer and use, Paint and varnish manufacturer, Resin manufacturer and se |
| For Use With (Equipment) | Half and Full Facemasks |
| Model | 6051 |
| Quantity | 1 Pair |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur