missing translation for 'onlineSavingsMsg'
Learn More

3M™ Gas and Vapour Filters

Product Code. 16802016
Click to view available options
Model:
6038
6051
6054
6055
6057
6059
6075
6099
Unit Size:
10 pairs
2 pairs
32 pairs
4 pairs

Product Code. 16802016

Brand: 3M™ 7000034749

Only null left
Add to basket
Add to basket

3M™ Gas and Vapour Filters; reusable masks with a bayonet connection and are designed to optimise your field of vision. Our filters offer lightweight and well-balanced breathing protection for hazardous environments.

Colour-coded labels according to the EN14387 classification system to show the type of protection they provide.These lightweight filters combine protection with comfort, and they’re compatible with our range of half and full-face masks for safe working in hazardous environments. They offer low breathing resistance and good balance when connected to your mask, and they’re designed to optimise your field of vision.They can be clicked into place quickly and easily, and they can be used with any 3M™ Reusable Masks that feature a bayonet-style connection. They can also be used in combination with 3M™ Particulate Filters and 3M™ 501 Particle Filter Retainers.
TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Cartridge Filter
Certifications/Compliance CE
Connection Type Bayonet
Description Organic Vapors with boiling point <65°C
Color White
Filter Type Gas and Vapor
Product Type Gas and Vapor Cartridge
Filter Efficiency A2
For Use With (Application) Protection against organic vapours (boiling point above 65%)
For Use With (Equipment) Half and Full Facemasks
Model 6055
Quantity 2/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.