Learn More
3M™ Gas & Vapor Filter GF22 A2
3M™ Gas & Vapor Filter DT-4001E is a DIN 40 threaded, replaceable filter offering A2 protection against certain organic vapors.
Brand: 3M™ 7100264834
93.70 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 2.00000kg
Description
- A2 protection against certain organic vapors
- DIN40 threaded connection
- Compatible with 3M™ Reusable Half Mask HF-300 Series, 3M™ Reusable Full Face Masks FF-300 Series and FF-600 Series
- Compatible with 3M™ Powered Air Respirator PF-600E Series
Specifications
Combination Cartridge/Filter | |
3M™ Reusable Half Mask HF-300 Series, Reusable Full Face Masks FF-300 Series, FF-600 Series, Powered Air Respirator PF-600E Series | |
DT-4001E | |
10/Cs. |