Learn More
3M™ Gas, Vapour and Particulate Filter
3M™ Gas, Vapour and Particulate Filters have a bayonet-style connection, and they’re designed to optimise your field of vision. Our lightweight filters offer well-balanced breathing protection for hazardous environments.
Brand: 3M™ 7100066103
3.20 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Additional Details : Weight : 0.04500kg
Description
Unique, compatible Bayonet Filter Attachment System allows different types of filters to just be clicked into place ensuring optimum ease of use.
- Lightweight, low breathing resistance, and well-balanced when fitted to mask
- Provides excellent field of vision as a result of the unique trapezoidal shape
- Bayonet-style cartridge connection for fitting to 3M 6000 and 7907 Series Full Face Masks
Specifications
Filter Retainer | |
Typical industrial applications for filters are laboratories and particulate applications | |
3M™ 6000 and 3M™ 7907 Series Full Face Masks | |
501 |