Learn More
Lenz Laborglasintrumente™ Gas Washing Bottle Head with Filter Disc
Gas washing head for bottles acc. to DIN 12463
Brand:  Lenz Laborglasintrumente™ 05510411
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11973551
  • £65.00 / Each
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12596
  • With cone NS 29/32
  • Material: DURAN™ tubing
Specifications
Specifications
Duran Tubing
1
1
GL 14 connections
Gas washing bottles acc. to DIN 12463