Learn More
Heidolph Instruments Gasket Seal for Rotary Evaporators
Made of glass materials, the ultra resistant FFKM Gasket Seal for applications with highly abrasive solutions is ideal in research, scientific, and other laboratory environments.
Brand:  Heidolph Instruments 23-30-01-01-30
178.06 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Product Code. 10592672
  • £193.00 / Each
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Vacuum seal
  • Standard FKM spare seal
  • Ultra resistant FFKM seal for applications with highly abrasive solutions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTFE
Gasket
115/230 VAC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Heidolph Instruments Gasket Seal for Rotary Evaporators > Gasket Seal for Rotary Evaporators; Material: Glass