missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gasoline Range Organics, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Descripción
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| 1,2,4-Trimethylbenzene | 95-63-6 | 1000μg/mL |
| 2,2,4-Trimethylpentane | 540-84-1 | 1000μg/mL |
| 3-Methylpentane | 96-14-0 | 1000μg/mL |
| Benzene | 71-43-2 | 1000μg/mL |
| Ethylbenzene | 100-41-4 | 1000μg/mL |
| m-Xylene | 108-38-3 | 1000μg/mL |
| Naphthalene | 91-20-3 | 1000μg/mL |
| o-Xylene | 95-47-6 | 1000μg/mL |
| Toluene | 108-88-3 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Especificaciones
Especificaciones
| Chemical Name or Material | Gasoline Range Organics |
| Packaging | 2mL amber prescored glass ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| Methods | For US EPA Method 8015 |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Solvent or Matrix | Methanol-P and T |
| Quantity | 1 mL |
Seguridad y manipulación
Nombre del producto
- GASOLINE RANGE ORGANICS
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 1A
- Flammable liquid Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : benzene, napthalene,ethylbenzene, toluene, methanol
Product intended for laboratory use only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido