missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Gast High-Capacity Vacuum Pump with Gauge, Regulator, and Relief
If you need a small, quiet source of vacuum or pressure, you’ll find the unit for your application in the Gast line High-Capacity Vacuum Pump. These pumps are available in single- and dual-head styles.
Brand: Cole-Parmer™ 07061-05
This item is not returnable.
View return policy
Description
- Free-air capacities up to 2.2 cfm (62.3 L/min.)
- Shaded-pole, fan-cooled motor has thermal overload switch to protect your pump system
- Different pump models have heads in parallel for higher capacity or series for greater vacuum
- Select vacuum pumps with or without gauges, regulators, and valves
- Runs dry
Specifications
1/4 hp | |
2 amp | |
68 dB | |
38°C | |
50 Hz | |
220 V |
Permanent Split Capacitor | |
High-Capacity Vacuum Pump | |
100°F | |
1.79 CFM, 51 L/min. | |
Vacuum System |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
- Multiple Manufacturers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Gast High-Capacity Vacuum Pump with Gauge, Regulator, and Relief > Vacuum Pump; Motor Type: Permanent Split Capacitor
Spot an opportunity for improvement?Share a Content Correction