Learn More
Abnova™ GATA3 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Brand:  Abnova™ H00002625-T01.100uL
Product Code. 16188544
  • £182.00 / 100µL
Explore more special offers
Specifications
293T
GATA3 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 48.95kDa
Transient overexpression cell lysate was tested with anti-GATA3 antibody (Mfr. No. H00002625-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
NM_001002295.1
pCMV-GATA3 full-length