Learn More
GATA6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540559
288.01 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Peptide sequence: PEEMYQTLAA LSSQGPAAYD GAPGGFVHSA AAAAAAAAAA SSPVYVPTTR Sequence homology: Cow: 92%; Dog: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%
GATA6 Polyclonal antibody specifically detects GATA6 in Human samples. It is validated for Western Blot.
Specifications
GATA6 | |
Polyclonal | |
Unconjugated | |
GATA6 | |
AA410133; ASD9; AVSD5; DNA-binding protein GATA-GT2; GATA binding protein 6; Gata6; GATA-6; GATA-binding factor 6; GATA-binding protein 6; LOW QUALITY PROTEIN: transcription factor GATA-6; transcription factor GATA6; transcription factor GATA-6; zinc-finger transcription factor | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2627 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q92908 | |
GATA6 | |
Synthetic peptide directed towards the N-terminal of human GATA6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.