missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gatifloxacin sesquihydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
A broad spectrum antibiotic
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Type | Gatifloxacin Sesquihydrate |
Physical Form | Powder |
Color | White to Yellow |
Quantity | 5 g |
Boiling Point | 684°C |
Merck Index | 14,4376 |
Solubility Information | Soluble in 1N NaOH at 10mg/mL |
Formula Weight | 402.42 |
Chemical Name or Material | Gatifloxacin sesquihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'