Learn More
Thermo Scientific™ Gator A2 Large System 24-Slot Rapid Load UVT Gel Trays, with gasketed End Gates
A2 and A5 Large Gel 24-slot Rapid Load UVT Gel Trays enable multiple sample loading.
Brand: Thermo Scientific™ A2-RL-UVT
Code : 04
Additional Details : Weight : 0.11100kg
Description
Thermo Scientific™Owl A2 and A5 Large Gel 24-slot Rapid Load UVT Gel Trays enable multiple sample loading.
Each side of UVT gel tray is fitted with a gasketed end gate which provides a leakproof seal without tape. Built in leveling ensures the casting of flat, uniformly thick gels.Specifications
24-slot rapid load UVT gel tray with gasketed end gates | |
A2 and A5 Large Gel Electrophoresis System | |
Gel Tray |