missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ pTK-Gaussia Luc Vector for Luciferase Assays
Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Thermo Scientific™ pMCS-Gaussia Luc Vector is a multiple cloning site plasmid designed to accept a promoter sequence for study of gene regulation using the naturally secreting Gaussia luciferase reporter.
The Gaussia Luc Vectors contain a gene cloned from the copepod, Gaussia princeps. The gene encodes a naturally-secreted bioluminescent Gaussia luciferase (approx. 20kDa), which enables measurement of the reporter activity in media (for real-time assays) and in cell lysates. The pMCS vector contains a multiple cloning site for cloning a promoter to study its regulatory potential. The pCMV and pTK vectors have the luciferase gene under the CMV (Cytomegalovirus) promoter and Herpes Simplex Virus (HSV) thymidine kinase (TK) promoter, respectively. These constitutive expression vectors can be used as normalization controls to account for experimental variation in combination with other reporters.
Highlights:
- Naturally-secreting Gaussia luciferase gene, optimized for high expression in mammalian systems
- Multiple cloning site (MCS) provides versatility for transfer of regulatory elements from one plasmid to another
- Transcription termination site (Ter), Lac operator (Lac O1), and transcriptional pause site (TPS) used to minimize background by reducing transcriptional read-through
- Both puromycin (Pur) and ampicillin (Amp) markers for drug selection in mammalian and bacterial cells, respectively
- High-copy pUC bacterial DNA replication origin
- Two control vectors available with strong (CMV) or weak (TK) constitutive promoters for co-transfection and normalization
Specifications
Specifications
| Description | pTK-Gaussia Luc Vector |
| Type | Luc Vector |
| Cloning Method | Restriction Enzyme/MCS |
| Delivery Type | Transfection |
| Quantity | 10 μg |
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: -70 to -20°C. |
| Reporter Gene | Gaussia Luciferase |
| Selection Agent (Eukaryotic) | Puromycin |
| Product Line | Pierce |
| For Use With (Application) | Reporter Assays |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction