Learn More
Lenz Laborglasintrumente™ Gay Lussac Pycnometers Unadjusted
Pycnometers acc. to Gay Lussac, unadjusted
Brand:  Lenz Laborglasintrumente™ 05630028
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10000281
  • £38.00 / Each
Estimated Shipment: 09-08-2024
to see stock.
Add to basket
Description
Description
  • Gay Lussac pycnometers acc. to DIN ISO 3507 and in addition to standard
  • Unadjusted
  • Stopper NS 10/19 with capillary
  • Stopper top ground and polished
  • Imprint of nominal capacity at the bottom
  • Material: borosilicate glass 3.3
Specifications
Specifications
1
50 mL
Borosilicate Glass (3.3)