missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBX1, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00002636-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MQRAGGGSAPGGNGGGGGGGPGTAFSIDSLIGPPPPRSGHLLYTGYPMFMPYRPLVLPQALAPAPLPAGLPPLAPLASFAGRLTNTFCAGLGQAVPSMVALTTALPSFAEPPDAFYGPQELAAAAAAAAATAARNNPEPGGRRPEGGLEADELLPAREKVAEPPPPPPPHFSETFPSLPAEGKVYSSDEEKLEASAGDPAGSEQEEEGSGGDSEDDGFLDSSAGGPGALLGPKPKLKGSLGTGAEEGAPVTAGVTAPGGKSRRRRTAFTSEQLLELEKEFHCKKYLSLTERSQIAHALKLSEVQVKIWFQNRRAKWKRIKAGNVSSRSGEPVRNPKIVVPIPVHVNRFAVRSQHQQMEQGARPSpecifications
GBX1 | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
BC160003.1 | |
GBX1 | |
GBX1 (AAI60003.1, 1 a.a. ∼ 363 a.a) full-length human protein. | |
RUO | |
2636 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
gastrulation brain homeobox 1 | |
- | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction