Learn More
Trajan™ BP624 GC Capillary Columns
SGE BP624 GC Capillary Columns - A polar phase designed for EPA volatile organics analysis
£555.00 - £1110.00
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||
12965571
|
Trajan™
054827 |
30 mm | 0.22 mm | 1.2 mm |
£575.00
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12760720
|
Trajan™
054836 |
30 mm | 0.53 mm | 3 mm |
£627.00
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12925581
|
Trajan™
054840 |
30 m | 0.25 mm | 1.4 μm |
£555.00
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12935581
|
Trajan™
054841 |
60 m | 0.32 mm | 1.8 μm |
£924.00
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
12985571
|
Trajan™
054832 |
30 m | 0.32 mm | 1.8 μm |
£591.00
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||
12915581
|
Trajan™
054835 |
50 m | 0.53 mm | 3 μm |
£1110.00
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
Description
- Polar phase
- EPA organics analysis (EPA 624, 502.2, SW-846, 8240/8260)
- Ideal for environmental analyses, volatile organics, alcohol analysis
Specifications
230/240°C |